Product Details
- SNP ID
-
rs7556606
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
42
- Location
-
Chr.1:27780548 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTCAGAAAGATCATGGACTCTCCG[A/G]GGTCAGCAGATCTGTGACTGAATCC
- Phenotype
-
MIM: 606892
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
STX12
PubMed Links
Gene Details
- Gene
- STX12
- Gene Name
- syntaxin 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177424.2 |
|
Intron |
|
|
NP_803173.1 |
View Full Product Details