Product Details

SNP ID
rs9906261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16621615 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATCTCACTGAGCATTTCAATTTG[A/C]ATTTCCCTGAGTGCTTTTTAGGTTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF624 PubMed Links

Gene Details

Gene
ZNF624
Gene Name
zinc finger protein 624
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020787.3 3363 UTR 3 NP_065838.2
XM_006721562.3 3363 UTR 3 XP_006721625.1
XM_011523970.2 3363 UTR 3 XP_011522272.1

View Full Product Details