Product Details

SNP ID
rs10034385
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:176684662 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCCATCCTAGACTTCCAGTTTC[C/T]AGTAGCGGTGAGAGGACAAATTATC
Phenotype
MIM: 601528
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
VEGFC PubMed Links
Additional Information
For this assay, SNP(s) [rs114475399] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
VEGFC
Gene Name
vascular endothelial growth factor C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005429.4 Intron NP_005420.1

View Full Product Details