Product Details
- SNP ID
-
rs7255856
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:11299392 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCCATCACACGTGCCACCTTTGAT[A/G]GTGTGAGAAGCTGAAATCCTCAGCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HSPC102
PubMed Links
Gene Details
- Gene
- HSPC102
- Gene Name
- uncharacterized HSPC102
There are no transcripts associated with this gene.
- Gene
- TSPAN16
- Gene Name
- tetraspanin 16
View Full Product Details