Product Details

SNP ID
rs10072995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:129755646 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTGATTGTTTCTTAATTTCTGCT[C/G]TGTCTCTTGTCAAGTCCCCTTTACA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIAA1024L PubMed Links

Gene Details

Gene
KIAA1024L
Gene Name
KIAA1024 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257308.1 Intron NP_001244237.1

View Full Product Details