Product Details

SNP ID
rs6026140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58237232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCCCGCCGGGCACCGAGCGCGC[C/T]TACTTCCCGTGCGCCGCCGCTAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD60 PubMed Links

Gene Details

Gene
ANKRD60
Gene Name
ankyrin repeat domain 60
There are no transcripts associated with this gene.

Gene
PPP4R1L
Gene Name
protein phosphatase 4 regulatory subunit 1 like (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details