Product Details
- SNP ID
-
rs5910808
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:120239338 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGGCAGGCAGGCACGAAAGAAAA[G/T]AAAAGAAAGAGAAGAGAAGAGAAGA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NKAPP1
PubMed Links
Gene Details
- Gene
- NKAPP1
- Gene Name
- NFKB activating protein pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details