Product Details

SNP ID
rs10209338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70981066 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGATCCCCAGGTGCTTCTAACTTG[A/C]AGCTAAGTTTGGGAACCTCTGTCTC
Phenotype
MIM: 617009
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD53 PubMed Links

Gene Details

Gene
ANKRD53
Gene Name
ankyrin repeat domain 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001115116.1 Intron NP_001108588.1
NM_024933.3 Intron NP_079209.3
XM_005264569.4 Intron XP_005264626.1
XM_011533114.2 Intron XP_011531416.1
Gene
TEX261
Gene Name
testis expressed 261
There are no transcripts associated with this gene.

View Full Product Details