Product Details

SNP ID
rs7515784
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:51289162 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTCCTGTCATGTCCTTAGTCCAC[C/T]GCCACCTAAGTCCCGACCTCCCTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTC39A PubMed Links

Gene Details

Gene
TTC39A
Gene Name
tetratricopeptide repeat domain 39A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080494.3 Intron NP_001073963.1
NM_001144832.2 Intron NP_001138304.1
NM_001297662.1 Intron NP_001284591.1
NM_001297663.1 Intron NP_001284592.1
NM_001297664.1 Intron NP_001284593.1
NM_001297665.1 Intron NP_001284594.1
NM_001297666.1 Intron NP_001284595.1
NM_001297667.1 Intron NP_001284596.1
XM_005270644.2 Intron XP_005270701.1
XM_006710471.2 Intron XP_006710534.1
XM_011541048.2 Intron XP_011539350.1
XM_011541049.2 Intron XP_011539351.1
XM_011541050.2 Intron XP_011539352.1
XM_011541051.2 Intron XP_011539353.1
XM_011541052.2 Intron XP_011539354.1
XM_011541053.2 Intron XP_011539355.1
XM_011541054.2 Intron XP_011539356.1
XM_017000715.1 Intron XP_016856204.1

View Full Product Details