Product Details

SNP ID
rs6070698
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:59033863 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTGTTTTTCAGTTTGCAGCTGCC[C/T]TCTTGACATCCCAAATAAGGAATTC
Phenotype
MIM: 606153 MIM: 612901
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP5E PubMed Links

Gene Details

Gene
ATP5E
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, epsilon subunit
There are no transcripts associated with this gene.

Gene
PRELID3B
Gene Name
PRELI domain containing 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256403.1 Intron NP_001243332.1
NM_016045.2 Intron NP_057129.2
Gene
SLMO2-ATP5E
Gene Name
SLMO2-ATP5E readthrough
There are no transcripts associated with this gene.

Gene
TUBB1
Gene Name
tubulin beta 1 class VI
There are no transcripts associated with this gene.

View Full Product Details