Product Details
- SNP ID
-
rs9924094
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:3961462 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACCCTGTCTCCAAAAAAAAAAAAA[A/C]CGAACAAAACCTCCATTCTTACAAG
- Phenotype
-
MIM: 603302
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ADCY9
PubMed Links
Gene Details
- Gene
- ADCY9
- Gene Name
- adenylate cyclase 9
View Full Product Details