Product Details

SNP ID
rs3211098
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:227329765 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTAAATGAGTAAAGGAACAAGCA[G/T]TGACACATCACTAGGAAGGTCAGTG
Phenotype
MIM: 614785
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
LOC654841 PubMed Links
Additional Information
For this assay, SNP(s) [rs3211097] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LOC654841
Gene Name
uncharacterized LOC654841
There are no transcripts associated with this gene.

Gene
MFF
Gene Name
mitochondrial fission factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277061.1 533 Intron NP_001263990.1
NM_001277062.1 533 Intron NP_001263991.1
NM_001277063.1 533 Intron NP_001263992.1
NM_001277064.1 533 Intron NP_001263993.1
NM_001277065.1 533 Intron NP_001263994.1
NM_001277066.1 533 Intron NP_001263995.1
NM_001277067.1 533 Intron NP_001263996.1
NM_001277068.1 533 Intron NP_001263997.1
NM_020194.5 533 Intron NP_064579.3
XM_005246681.3 533 Intron XP_005246738.1
XM_005246682.1 533 Intron XP_005246739.1
XM_005246683.1 533 Intron XP_005246740.1
XM_005246684.1 533 Intron XP_005246741.1
XM_005246685.3 533 Intron XP_005246742.1
XM_005246686.3 533 Intron XP_005246743.1
XM_005246687.3 533 Intron XP_005246744.1
XM_005246688.1 533 Intron XP_005246745.1
XM_006712637.1 533 Intron XP_006712700.1
XM_006712638.1 533 Intron XP_006712701.1
XM_006712639.2 533 Intron XP_006712702.1
XM_011511500.1 533 Intron XP_011509802.1
XM_011511501.1 533 Intron XP_011509803.1
XM_017004512.1 533 Missense Mutation AGT,ATT S7I XP_016860001.1
XM_017004513.1 533 Intron XP_016860002.1
XM_017004514.1 533 Intron XP_016860003.1
XM_017004515.1 533 Intron XP_016860004.1

View Full Product Details