Product Details
- SNP ID
-
rs9431947
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
41
- Location
-
Chr.1:231166610 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTCATTGCATCACCAGAACTAGAA[C/T]TGGGACAGCAACATTTCTCTAACAT
- Phenotype
-
MIM: 610584
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TRIM67
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114929190] are located under a probe and SNP(s) [rs115461650] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TRIM67
- Gene Name
- tripartite motif containing 67
View Full Product Details