Product Details
- SNP ID
-
rs6004371
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:24964790 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGAGGAAACTGAGTTTTGGAGAA[A/G]GAGTCACTTGCCCAGGACCATACTA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1671
PubMed Links
Gene Details
- Gene
- KIAA1671
- Gene Name
- KIAA1671
View Full Product Details