Product Details

SNP ID
rs9973862
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201800336 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAAAATAGCCATGAATATAAAAA[A/C]AGAAGTACATTATTGAAATGTATAG
Phenotype
MIM: 616147
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CDK15 PubMed Links

Gene Details

Gene
CDK15
Gene Name
cyclin dependent kinase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261435.1 Intron NP_001248364.1
NM_001261436.1 Intron NP_001248365.1
NM_139158.2 Intron NP_631897.1
XM_005246781.3 Intron XP_005246838.1
XM_005246782.4 Intron XP_005246839.1
XM_011511650.2 Intron XP_011509952.1
XM_011511652.1 Intron XP_011509954.1
XM_011511654.2 Intron XP_011509956.1
XM_011511655.2 Intron XP_011509957.1
XM_011511656.2 Intron XP_011509958.1
XM_011511657.2 Intron XP_011509959.1

View Full Product Details