Product Details

SNP ID
rs9912517
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81988497 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGAGCAAGTGGGGCAGCTCTTTC[A/G]AGACACCAGGGACAGCAGGAGGTGG
Phenotype
MIM: 606236
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ASPSCR1 PubMed Links
Additional Information
For this assay, SNP(s) [rs143827048] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ASPSCR1
Gene Name
ASPSCR1, UBX domain containing tether for SLC2A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251888.1 Intron NP_001238817.1
NM_024083.3 Intron NP_076988.1
XM_011523601.2 Intron XP_011521903.1
XM_011523602.2 Intron XP_011521904.1
XM_017025036.1 Intron XP_016880525.1
XM_017025037.1 Intron XP_016880526.1
XM_017025038.1 Intron XP_016880527.1
XM_017025039.1 Intron XP_016880528.1
XM_017025040.1 Intron XP_016880529.1
XM_017025041.1 Intron XP_016880530.1
XM_017025042.1 Intron XP_016880531.1
XM_017025043.1 Intron XP_016880532.1
XM_017025044.1 Intron XP_016880533.1

View Full Product Details