Product Details

SNP ID
rs9818363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122393444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTTCTTTCAGCTTCACAATAG[C/T]CTTGAAAACCAACAGCCTCACAAAT
Phenotype
MIM: 608017
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM162A PubMed Links

Gene Details

Gene
FAM162A
Gene Name
family with sequence similarity 162 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014367.3 Intron NP_055182.3

View Full Product Details