Product Details

SNP ID
rs9611424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40830659 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATAAAAGCTCACTGAATAATTTA[A/C]GACTAAAAATGTCCCCCAAATTAGA
Phenotype
MIM: 606796
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ST13 PubMed Links

Gene Details

Gene
ST13
Gene Name
suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278589.1 Intron NP_001265518.1
NM_003932.4 Intron NP_003923.2

View Full Product Details