Product Details

SNP ID
rs9394121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32878207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTATTTTTTAAAAAACTGAAGT[C/T]TGCCCAGAGTACCAAAAACATTAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1R2P1 PubMed Links

Gene Details

Gene
PPP1R2P1
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 2 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details