Product Details

SNP ID
rs6116498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:4724227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACTTGGGAGTATCAAAGAAAAC[C/T]TACATGTATTAAGTCACCACTCCTT
Phenotype
MIM: 604263
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRND PubMed Links

Gene Details

Gene
PRND
Gene Name
prion protein 2 (dublet)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012409.3 Intron NP_036541.2
Gene
PRNT
Gene Name
prion protein (testis specific)
There are no transcripts associated with this gene.

View Full Product Details