Product Details

SNP ID
rs10452640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:121437338 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAGTTGTCAATCTAATTTCCCAT[A/C]CCTCTCACACCAGCCCTTTTTTTTT
Phenotype
MIM: 121014
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GJA1 PubMed Links

Gene Details

Gene
GJA1
Gene Name
gap junction protein alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000165.4 Intron NP_000156.1

View Full Product Details