Product Details

SNP ID
rs10500543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68247408 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGGCTGTCCCCTCCCAGTGTTAC[C/T]TGAGGTGAAAGGCATGTGGATGGCT
Phenotype
MIM: 609362
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLA2G15 PubMed Links

Gene Details

Gene
PLA2G15
Gene Name
phospholipase A2 group XV
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012320.3 Intron NP_036452.1
XM_005255886.3 Intron XP_005255943.1
XM_011522979.2 Intron XP_011521281.1
XM_011522980.2 Intron XP_011521282.2

View Full Product Details