Product Details

SNP ID
rs6131418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1370664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTTCCAGACTCTTGCAAGAAG[C/T]TGAGACATTCTTTGGCAGTGGGTTG
Phenotype
MIM: 186945
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FKBP1A PubMed Links

Gene Details

Gene
FKBP1A
Gene Name
FK506 binding protein 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000801.4 Intron NP_000792.1
NM_001199786.1 Intron NP_001186715.1
NM_054014.3 Intron NP_463460.1
Gene
FKBP1A-SDCBP2
Gene Name
FKBP1A-SDCBP2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
SDCBP2-AS1
Gene Name
SDCBP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details