Product Details

SNP ID
rs13062222
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:47808548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATTTCGTAGCAATAGAGTATTG[G/T]TTCTTTTTTTTTTTTCTTAAATTTA
Phenotype
MIM: 616423
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
DHX30 PubMed Links

Gene Details

Gene
DHX30
Gene Name
DEAH-box helicase 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014966.3 Intron NP_055781.2
NM_138615.2 Intron NP_619520.1
XM_006713033.1 Intron XP_006713096.1
XM_011533490.1 Intron XP_011531792.1
XM_011533491.1 Intron XP_011531793.1
XM_011533492.2 Intron XP_011531794.1
XM_011533493.1 Intron XP_011531795.1
XM_011533494.2 Intron XP_011531796.1
XM_011533495.1 Intron XP_011531797.1
XM_011533496.1 Intron XP_011531798.1
XM_011533497.2 Intron XP_011531799.1
XM_011533498.1 Intron XP_011531800.1
XM_017005914.1 Intron XP_016861403.1
XM_017005915.1 Intron XP_016861404.1
XM_017005916.1 Intron XP_016861405.1
XM_017005917.1 Intron XP_016861406.1

View Full Product Details