Product Details

SNP ID
rs4393865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:28326465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTTTGGTGTATTCTGTAGCACT[A/G]GAATGAAAATTTGCTTTTGGTGTCT
Phenotype
MIM: 609916 MIM: 615166
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AZI2 PubMed Links

Gene Details

Gene
AZI2
Gene Name
5-azacytidine induced 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134432.1 Intron NP_001127904.1
NM_001134433.1 Intron NP_001127905.1
NM_001271650.1 Intron NP_001258579.1
NM_022461.4 Intron NP_071906.1
XM_005265392.2 Intron XP_005265449.1
XM_011534009.2 Intron XP_011532311.1
XM_011534010.2 Intron XP_011532312.1
XM_011534011.2 Intron XP_011532313.1
Gene
CMC1
Gene Name
C-X9-C motif containing 1
There are no transcripts associated with this gene.

View Full Product Details