Product Details

SNP ID
rs12089667
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:38844447 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTGGCTCTGACAAACAGGACTGA[A/G]GAAAAAAAAAAAAAAAACAGAAGAA
Phenotype
MIM: 608267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RRAGC PubMed Links

Gene Details

Gene
RRAGC
Gene Name
Ras related GTP binding C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271851.1 Intron NP_001258780.1
NM_022157.3 Intron NP_071440.1

View Full Product Details