Product Details

SNP ID
rs12125560
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:44065815 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGATATTTATAACATATATAATCA[A/G]CAAACAGTGTCTAGTATGTGCAGAT
Phenotype
MIM: 609602
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLF17 PubMed Links

Gene Details

Gene
KLF17
Gene Name
Kruppel like factor 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173484.3 Intron NP_775755.3
XM_005270465.3 Intron XP_005270522.2
XM_011540700.1 Intron XP_011539002.1
XM_011540704.2 Intron XP_011539006.1

View Full Product Details