Product Details
- SNP ID
-
rs12104154
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:2097833 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACCAGGTGAGCCTGGGCCCCAGTGC[C/T]GAGGGACAGGCCCTGAGGCTTTGGA
- Phenotype
-
MIM: 607246
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AP3D1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112353711] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AP3D1
- Gene Name
- adaptor related protein complex 3 delta 1 subunit
There are no transcripts associated with this gene.
- Gene
- IZUMO4
- Gene Name
- IZUMO family member 4
- Gene
- MOB3A
- Gene Name
- MOB kinase activator 3A
There are no transcripts associated with this gene.
View Full Product Details