Product Details

SNP ID
rs11202285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:86983934 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAGAAGTATAGCATTGCTCAAAT[A/G]ACCTGTGGTAAGTGTATTTATCACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGAP11 PubMed Links

Gene Details

Gene
AGAP11
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133447.1 357 UTR 5 NP_597704.1

View Full Product Details