Product Details

SNP ID
rs11978345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99332413 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGGTGATTACGTGGTGATTGCCA[G/T]ATTTCTTCAACGTAAAGGCATCTTT
Phenotype
MIM: 604220
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARPC1A PubMed Links

Gene Details

Gene
ARPC1A
Gene Name
actin related protein 2/3 complex subunit 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190996.1 Intron NP_001177925.1
NM_006409.3 Intron NP_006400.2

View Full Product Details