Product Details

SNP ID
hCV30926050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53532167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAGGCTGGAACCATCGAAGTCC[C/T]TAGATCAGCTTTAAGCACTGGATGG
Phenotype
MIM: 300697
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HUWE1 PubMed Links

Gene Details

Gene
HUWE1
Gene Name
HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
There are no transcripts associated with this gene.

View Full Product Details