Product Details
- SNP ID
-
rs6902316
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:160472135 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAGTTTTTTTGTGATATTCATCAC[A/G]TTAATGATAAAAATACATTTTAATA
- Phenotype
-
MIM: 611682
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LPAL2
PubMed Links
Gene Details
- Gene
- LPAL2
- Gene Name
- lipoprotein(a) like 2, pseudogene
There are no transcripts associated with this gene.
View Full Product Details