Product Details

SNP ID
rs12207325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:160482879 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCTAGAACAGAGAAACAGGTGAA[A/G]CCATTTATGACACAAACCAGGCAGG
Phenotype
MIM: 611682
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LPAL2 PubMed Links

Gene Details

Gene
LPAL2
Gene Name
lipoprotein(a) like 2, pseudogene
There are no transcripts associated with this gene.

View Full Product Details