Product Details

SNP ID
rs10967806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:27245756 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGGCTTTTGCATTTTCACTAGCA[C/T]TGTGACCTCCAAATAGTACAGAATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00032 PubMed Links

Gene Details

Gene
LINC00032
Gene Name
long intergenic non-protein coding RNA 32
There are no transcripts associated with this gene.

View Full Product Details