Product Details

SNP ID
rs13300876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97504637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGGTAAAACGAAGCAGATCCTG[A/G]CTCAGACCCCTATGTCTCTGGCAGA
Phenotype
MIM: 611258 MIM: 190930
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TDRD7 PubMed Links

Gene Details

Gene
TDRD7
Gene Name
tudor domain containing 7
There are no transcripts associated with this gene.

Gene
TMOD1
Gene Name
tropomodulin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166116.1 Intron NP_001159588.1
NM_003275.3 Intron NP_003266.1

View Full Product Details