Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001144763.1 | 6696 | UTR 3 | NP_001138235.1 | ||
NM_001144764.1 | 6696 | UTR 3 | NP_001138236.1 | ||
NM_001144765.1 | 6696 | UTR 3 | NP_001138237.1 | ||
NM_001308019.1 | 6696 | UTR 3 | NP_001294948.1 | ||
NM_015065.2 | 6696 | UTR 3 | NP_055880.2 | ||
XM_011542696.2 | 6696 | UTR 3 | XP_011540998.1 | ||
XM_011542698.2 | 6696 | UTR 3 | XP_011541000.1 | ||
XM_011542700.2 | 6696 | UTR 3 | XP_011541002.1 | ||
XM_017017397.1 | 6696 | UTR 3 | XP_016872886.1 | ||
XM_017017398.1 | 6696 | UTR 3 | XP_016872887.1 | ||
XM_017017399.1 | 6696 | UTR 3 | XP_016872888.1 | ||
XM_017017400.1 | 6696 | UTR 3 | XP_016872889.1 | ||
XM_017017401.1 | 6696 | UTR 3 | XP_016872890.1 | ||
XM_017017402.1 | 6696 | UTR 3 | XP_016872891.1 | ||
XM_017017403.1 | 6696 | UTR 3 | XP_016872892.1 | ||
XM_017017404.1 | 6696 | UTR 3 | XP_016872893.1 |