Product Details

SNP ID
rs11161465
Assay Type
Functionally tested
NCBI dbSNP Submissions
41
Location
Chr.1:75726059 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATAGCAACCTTTGCACTTTGTTC[C/T]ATGTAACATATTTTCCTGAACAGTC
Phenotype
MIM: 607008
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACADM PubMed Links

Gene Details

Gene
ACADM
Gene Name
acyl-CoA dehydrogenase, C-4 to C-12 straight chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000016.5 Intron NP_000007.1
NM_001127328.2 Intron NP_001120800.1
NM_001286042.1 Intron NP_001272971.1
NM_001286043.1 Intron NP_001272972.1
NM_001286044.1 Intron NP_001272973.1
Gene
SLC44A5
Gene Name
solute carrier family 44 member 5
There are no transcripts associated with this gene.

View Full Product Details