Product Details

SNP ID
rs11145228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:69102586 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTGCATTTCATTTTTGCATCGTG[A/G]TAATCATACTGGGTGAATTTTGTAT
Phenotype
MIM: 606829 MIM: 607709
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FXN PubMed Links

Gene Details

Gene
FXN
Gene Name
frataxin
There are no transcripts associated with this gene.

Gene
TJP2
Gene Name
tight junction protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170414.2 Intron NP_001163885.1
NM_001170415.1 Intron NP_001163886.1
NM_001170416.1 Intron NP_001163887.1
NM_004817.3 Intron NP_004808.2
NM_201629.3 Intron NP_963923.1
XM_005252314.2 Intron XP_005252371.1
XM_011519204.1 Intron XP_011517506.1
XM_011519206.2 Intron XP_011517508.1
XM_011519207.2 Intron XP_011517509.1
XM_011519208.2 Intron XP_011517510.1
XM_011519209.2 Intron XP_011517511.1
XM_017015326.1 Intron XP_016870815.1
XM_017015327.1 Intron XP_016870816.1
XM_017015328.1 Intron XP_016870817.1

View Full Product Details