Product Details

SNP ID
rs11145325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:69118192 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTAAACAGTAGCATCCTACATAC[A/G]CTTTTATTACTTTGTTAATACCCTA
Phenotype
MIM: 607709
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TJP2 PubMed Links

Gene Details

Gene
TJP2
Gene Name
tight junction protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170414.2 Intron NP_001163885.1
NM_001170415.1 Intron NP_001163886.1
NM_001170416.1 Intron NP_001163887.1
NM_004817.3 Intron NP_004808.2
NM_201629.3 Intron NP_963923.1
XM_005252314.2 Intron XP_005252371.1
XM_011519204.1 Intron XP_011517506.1
XM_011519206.2 Intron XP_011517508.1
XM_011519207.2 Intron XP_011517509.1
XM_011519208.2 Intron XP_011517510.1
XM_011519209.2 Intron XP_011517511.1
XM_017015326.1 Intron XP_016870815.1
XM_017015327.1 Intron XP_016870816.1
XM_017015328.1 Intron XP_016870817.1

View Full Product Details