Product Details

SNP ID
rs12158059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40769942 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCAGATGTTTATTTTCAAAATATA[C/T]GGCCTACACCAAGTTTATAAAGGGA
Phenotype
MIM: 606795
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A17 PubMed Links

Gene Details

Gene
SLC25A17
Gene Name
solute carrier family 25 member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282726.1 2020 UTR 3 NP_001269655.1
NM_001282727.1 2020 UTR 3 NP_001269656.1
NM_006358.3 2020 UTR 3 NP_006349.1

View Full Product Details