Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001251977.1 | 89 | Intron | NP_001238906.1 | ||
NM_001251978.1 | 89 | Intron | NP_001238907.1 | ||
NM_001251979.1 | 89 | UTR 5 | NP_001238908.1 | ||
NM_001251980.1 | 89 | Intron | NP_001238909.1 | ||
NM_001251981.1 | 89 | Intron | NP_001238910.1 | ||
NM_001251982.1 | 89 | Intron | NP_001238911.1 | ||
NM_001251983.1 | 89 | Intron | NP_001238912.1 | ||
NM_001251984.1 | 89 | UTR 5 | NP_001238913.1 | ||
NM_001251985.1 | 89 | Intron | NP_001238914.1 | ||
NM_013441.3 | 89 | Intron | NP_038469.1 |