Product Details

SNP ID
rs3101469
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:244410343 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCACACACTTCACTTTTTCTTAT[C/G]TTTCAAATATCCATCAATGCCAACA
Phenotype
MIM: 103060
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADSS PubMed Links

Gene Details

Gene
ADSS
Gene Name
adenylosuccinate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126.3 Intron NP_001117.2
XM_011544108.2 Intron XP_011542410.1

View Full Product Details