Product Details

SNP ID
rs12032765
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:244414592 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTTTCCTGCTGTGATAGCCACAA[C/T]AGGGGCTAATTTACTCCACTAACCT
Phenotype
MIM: 103060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADSS PubMed Links

Gene Details

Gene
ADSS
Gene Name
adenylosuccinate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126.3 Intron NP_001117.2
XM_011544108.2 Intron XP_011542410.1

View Full Product Details