Product Details

SNP ID
hCV31529506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29997145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGGGAAGAATAAGTAAATGAAG[A/C]CAATAGACTAGAATTTTAGGTCATA
Phenotype
MIM: 604567 MIM: 603365 MIM: 613199
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DOC2A PubMed Links

Gene Details

Gene
DOC2A
Gene Name
double C2 domain alpha
There are no transcripts associated with this gene.

Gene
HIRIP3
Gene Name
HIRA interacting protein 3
There are no transcripts associated with this gene.

Gene
INO80E
Gene Name
INO80 complex subunit E
There are no transcripts associated with this gene.

Gene
TAOK2
Gene Name
TAO kinase 2
There are no transcripts associated with this gene.

View Full Product Details