Product Details
- SNP ID
-
rs11052664
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:33379754 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAAGGGTGAATACATAAAATATCA[A/G]ATAAAGGTTAGCAAATAAATAATAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SYT10
PubMed Links
Gene Details
- Gene
- SYT10
- Gene Name
- synaptotagmin 10
View Full Product Details