Product Details

SNP ID
rs10747763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56209282 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCTATATTCACAGCCCCAGCTAT[C/G]AACATTCTTTTTATTTTTTAGTTAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RNF41 PubMed Links

Gene Details

Gene
RNF41
Gene Name
ring finger protein 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242826.1 Intron NP_001229755.1
NM_005785.3 Intron NP_005776.1
NM_194358.2 Intron NP_919339.1
NM_194359.2 Intron NP_919340.1
XM_005268561.4 Intron XP_005268618.1
XM_011537734.2 Intron XP_011536036.1
XM_011537735.2 Intron XP_011536037.1

View Full Product Details