Product Details

SNP ID
rs12303143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:104218551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTACATTTACTTTTTGAAACTTAA[A/G]TTTAGGCATATTTTAAGCATTAATC
Phenotype
MIM: 601112
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TXNRD1 PubMed Links

Gene Details

Gene
TXNRD1
Gene Name
thioredoxin reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001093771.2 Intron NP_001087240.1
NM_001261445.1 Intron NP_001248374.1
NM_001261446.1 Intron NP_001248375.1
NM_003330.3 Intron NP_003321.3
NM_182729.2 Intron NP_877393.1
NM_182742.2 Intron NP_877419.1
NM_182743.2 Intron NP_877420.1

View Full Product Details