Product Details

SNP ID
rs7404843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:15436851 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATGCATCATTTCATTTAGTCCCC[G/T]CAACAACCTCCTGCAGTACTAAGAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C16orf45 PubMed Links

Gene Details

Gene
C16orf45
Gene Name
chromosome 16 open reading frame 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142469.1 Intron NP_001135941.1
NM_033201.2 Intron NP_149978.1
XM_011522731.2 Intron XP_011521033.1
XM_017023824.1 Intron XP_016879313.1

View Full Product Details