Product Details

SNP ID
rs7108844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77620052 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTCACAAAGTGAAAAGACAAAAA[C/T]CAGTACAGCAAGCTTTTTGCTTATT
Phenotype
MIM: 609914 MIM: 602158
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AQP11 PubMed Links

Gene Details

Gene
AQP11
Gene Name
aquaporin 11
There are no transcripts associated with this gene.

Gene
CLNS1A
Gene Name
chloride nucleotide-sensitive channel 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293.2 Intron NP_001284.1
NM_001311199.1 Intron NP_001298128.1
NM_001311200.1 Intron NP_001298129.1
NM_001311201.1 Intron NP_001298130.1
NM_001311202.1 Intron NP_001298131.1
Gene
LOC107987163
Gene Name
uncharacterized LOC107987163
There are no transcripts associated with this gene.

View Full Product Details