Product Details

SNP ID
rs12913288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:44962021 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCACAGATTAAGAAATTTAAAGGG[C/T]ATGCTGTTATTGAATATTACTTGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TERB2 PubMed Links

Gene Details

Gene
TERB2
Gene Name
telomere repeat binding bouquet formation protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152448.2 Intron NP_689661.1
XM_011521240.2 Intron XP_011519542.1

View Full Product Details